CAS RN 207801-31-8
IUPAC Name:
zinc(2+) ditrifluoroacetate
Synonyms:
zinc(2+) ditrifluoroacetate
Molecular Weight (g/mol):
291.41
Molecular Formula:
C4F6O4Zn
InChi Key:
VCQWRGCXUWPSGY-UHFFFAOYSA-L
SMILES:
[Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
1
–
2
of
2
results
1
Zinc trifluoroacetate hydrate
CAS: 207801-31-8 Molecular Formula: C4F6O4Zn Molecular Weight (g/mol): 291.41 MDL Number: MFCD00061595 InChI Key: VCQWRGCXUWPSGY-UHFFFAOYSA-L Synonym: zinc trifluoroacetate hydrate,acmc-1ch94,zinc trifluoroacetate-water 1/2/1,zinc 2+ hydrate ditrifluoroacetate PubChem CID: 57375648 IUPAC Name: zinc;2,2,2-trifluoroacetate;hydrate SMILES: [Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
2
Sigma Aldrich Zinc trifluoroacetate hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.