accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOONSOOFFFFFFSOONSOOFFFMn

CAS RN 207861-55-0

IUPAC Name: manganese(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: manganese(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol): 615.21
Molecular Formula: C4F12MnN2O8S4
InChi Key: NNWDGVNRIAAYMY-UHFFFAOYSA-N
SMILES: [Mn++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)