accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207861-61-8

IUPAC Name: λ²-cobalt(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: λ²-cobalt(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol): 619.2
Molecular Formula: C4CoF12N2O8S4
InChi Key: HRURXKIZWNSHQB-UHFFFAOYSA-N
SMILES: [Co++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results