accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOONSOOFFFFFFSOONSOOFFFNi

CAS RN 207861-63-0

IUPAC Name: nickel(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: nickel(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol): 618.96
Molecular Formula: C4F12N2NiO8S4
InChi Key: VFUSIQAHVPEYFD-UHFFFAOYSA-N
SMILES: [Ni++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results