accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207974-17-2

FNCSF

CAS RN 207974-17-2

IUPAC Name: 1,3-difluoro-2-isothiocyanatobenzene
Synonyms: 1,3-difluoro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 171.16
Molecular Formula: C7H3F2NS
InChi Key: DBSXNGIBAKYMSS-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results