accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OBr

CAS RN 207986-25-2

IUPAC Name: 2-bromo-1-[4-(diethylamino)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(diethylamino)phenyl]ethanone
Molecular Weight (g/mol): 270.17
Molecular Formula: C12H16BrNO
InChi Key: ALTMCDJIFXTEPV-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results