accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2080-75-3

IUPAC Name: 5-methoxy-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5-methoxy-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: AUPLVAKFTYFHTA-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results