accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNCl

CAS RN 208167-83-3

IUPAC Name: tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 248.75
Molecular Formula: C11H21ClN2O2
InChi Key: MYOWELLYEZMECA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CCCl)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)