accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 208173-24-4

IUPAC Name: 1-[4-fluoro-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-fluoro-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 206.14
Molecular Formula: C9H6F4O
InChi Key: SYFHRXQPXHETEF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results