accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2082-76-0

IUPAC Name: decanoyl decanoate
Synonyms: decanoic anhydride
Molecular Weight (g/mol): 326.52
Molecular Formula: C20H38O3
InChi Key: HTWWKYKIBSHDPC-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)