accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHCH3H3CCH3CH3CH3H3C

CAS RN 2082-79-3

IUPAC Name: octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Synonyms: octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 530.88
Molecular Formula: C35H62O3
InChi Key: SSDSCDGVMJFTEQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results