accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20821-66-3

IUPAC Name: N,N-dimethyl(prop-2-en-1-ylsulfanyl)carbothioamide
Synonyms: N,N-dimethyl(prop-2-en-1-ylsulfanyl)carbothioamide
Molecular Weight (g/mol): 161.28
Molecular Formula: C6H11NS2
InChi Key: ZFMLVJCASVPGIT-UHFFFAOYSA-N
SMILES: CN(C)C(=S)SCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results