accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)O

CAS RN 20826-94-2

IUPAC Name: ethyl 2-[(1R)-2-oxocyclopentyl]acetate
Synonyms: ethyl 2-[(1R)-2-oxocyclopentyl]acetate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: PJMKFKUFBDXYEC-SSDOTTSWSA-N
SMILES: CCOC(=O)C[C@H]1CCCC1=O
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)