CAS RN 208260-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ ZM 336372
A potent, cell-permeable, reversible, ATP-competitive and specific inhibitor of the protein kinase c-Raf (IC50 = 70 nM).