accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3HS

CAS RN 2084-18-6

IUPAC Name: 3-methylbutane-2-thiol
Synonyms: 3-methylbutane-2-thiol
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: BFLXFRNPNMTTAA-UHFFFAOYNA-N
SMILES: CC(C)C(C)S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results