accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20849-78-9

IUPAC Name: 4-(2-chloroethyl)benzoic acid
Synonyms: 4-(2-chloroethyl)benzoic acid
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: OOAPBGPLZAFZSO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CCCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results