accessibility menu, dialog, popup

UserName

Viewing profile as user:
OAlONONN

CAS RN 2085-33-8

IUPAC Name: 8-{[bis(quinolin-8-yloxy)alumanyl]oxy}quinoline
Synonyms: 8-{[bis(quinolin-8-yloxy)alumanyl]oxy}quinoline
Molecular Weight (g/mol): 459.44
Molecular Formula: C27H18AlN3O3
InChi Key: TVIVIEFSHFOWTE-UHFFFAOYSA-K
SMILES: O([Al](OC1=C2N=CC=CC2=CC=C1)OC1=C2N=CC=CC2=CC=C1)C1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (4)
  • (1)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (5)
  • (4)
  • (3)
  • (4)
  • (3)