accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2086-26-2

IUPAC Name: hydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride
Synonyms: hydrogen O-[(4-nitrophenyl)methyl]hydroxylamine chloride
Molecular Weight (g/mol): 204.61
Molecular Formula: C7H9ClN2O3
InChi Key: LKCAFSOYOMFQSL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)