CAS RN 20873-58-9
IUPAC Name:
5-(methylsulfanyl)thiophene-2-carboxylic acid
Synonyms:
5-(methylsulfanyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol):
174.23
Molecular Formula:
C6H6O2S2
InChi Key:
FWFWUSLBIIIIEN-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(S1)C(O)=O
1
–
2
of
2
results
2
Promotions Available
5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals