accessibility menu, dialog, popup

UserName

Viewing profile as user:
NS

CAS RN 20893-30-5

IUPAC Name: 2-(thiophen-2-yl)acetonitrile
Synonyms: 2-thiopheneacetonitrile
Molecular Weight (g/mol): 123.17
Molecular Formula: C6H5NS
InChi Key: CLSHQIDDCJTHAJ-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CS1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)