CAS RN 2092-65-1
IUPAC Name:
[({[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
Synonyms:
adenosine diphosphate
Molecular Weight (g/mol):
427.2
Molecular Formula:
C10H15N5O10P2
InChi Key:
XTWYTFMLZFPYCI-UHFFFAOYNA-N
SMILES:
NC1=C2N=CN(C3OC(COP(O)(=O)OP(O)(O)=O)C(O)C3O)C2=NC=N1
1
–
1
of
1
results
1
Adenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals
CAS: 2092-65-1 Molecular Formula: C10H15N5Na3O10P2 Molecular Weight (g/mol): 496.172 InChI Key: VJZLETZZUNQROW-MSQVLRTGSA-N Synonym: adenosine-5'-diphosphate trisodium salt PubChem CID: 131875859 IUPAC Name: [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium SMILES: C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na]