accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20925-60-4

IUPAC Name: 4-chloro-1H-indazol-3-amine
Synonyms: 4-chloro-1H-indazol-3-amine
Molecular Weight (g/mol): 167.6
Molecular Formula: C7H6ClN3
InChi Key: QPLXQNVPEHUPTR-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=CC(Cl)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results