accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFOH

CAS RN 209328-55-2

IUPAC Name: 5-fluoropyridin-3-ol
Synonyms: 5-fluoropyridin-3-ol
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: IUFLFKASIHPKNZ-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results