accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 209395-32-4

IUPAC Name: 1-(prop-2-yn-1-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(prop-2-yn-1-yl)pyrrole-2,5-dione
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: OBYJFWVFCFYKNY-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CC#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results