accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONH3CH3CFHOOH

CAS RN 2096333-31-0

IUPAC Name: {4-[ethyl(methyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 4-[ethyl(methyl)carbamoyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 225.03
Molecular Formula: C10H13BFNO3
InChi Key: UJSORFNSZVADTI-UHFFFAOYSA-N
SMILES: CCN(C)C(=O)C1=CC(F)=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results