CAS RN 2096336-05-7
IUPAC Name:
{4-[methyl(phenyl)sulfamoyl]phenyl}boronic acid
Synonyms:
4-[methyl(phenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol):
291.13
Molecular Formula:
C13H14BNO4S
InChi Key:
PUWVRBKKJNKHLI-UHFFFAOYSA-N
SMILES:
CN(C1=CC=CC=C1)S(=O)(=O)C1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
4-[Methyl(phenyl)sulfamoyl]benzeneboronic acid, 97%, Thermo Scientific™
CAS: 2096336-05-7 Molecular Formula: C13H14BNO4S Molecular Weight (g/mol): 291.13 MDL Number: MFCD20265341 InChI Key: PUWVRBKKJNKHLI-UHFFFAOYSA-N Synonym: 4-methyl phenyl sulfamoyl benzeneboronic acid PubChem CID: 73995963