accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSOONHFHOOH

CAS RN 2096336-84-2

IUPAC Name: {3-[(3-fluorophenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[(3-fluorophenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 295.09
Molecular Formula: C12H11BFNO4S
InChi Key: SFLKIWAVAZHFPE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)NC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results