accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2096340-10-0

IUPAC Name: (3-{[2-(ethylamino)ethyl]carbamoyl}-5-fluorophenyl)boronic acid
Synonyms: 3-{[2-(ethylamino)ethyl]carbamoyl}-5-fluorophenylboronic acid
Molecular Weight (g/mol): 254.07
Molecular Formula: C11H16BFN2O3
InChi Key: BGUHDVCROSGNPJ-UHFFFAOYSA-N
SMILES: CCNCCNC(=O)C1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results