accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 20965-27-9

IUPAC Name: 7-bromoheptanenitrile
Synonyms: 7-bromoheptanenitrile
Molecular Weight (g/mol): 190.08
Molecular Formula: C7H12BrN
InChi Key: HVVQSKCGHAPHMV-UHFFFAOYSA-N
SMILES: BrCCCCCCC#N
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)