accessibility menu, dialog, popup

UserName

Viewing profile as user:
BClCH3HOOH

CAS RN 209919-30-2

IUPAC Name: (4-chloro-2-methylphenyl)boronic acid
Synonyms: 4-chloro-2-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: SRXXSLUUAWHGBZ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Cl)=C1)B(O)O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results