accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNN

CAS RN 20996-87-6

IUPAC Name: 1-methyl-1H-indole-6-carbonitrile
Synonyms: 1-methylindole-6-carbonitrile
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: FOGAAHLMYPQCJG-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=C(C=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results