accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 209980-53-0

IUPAC Name: N4,N4'-bis[4-(diphenylamino)phenyl]-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis[4-(diphenylamino)phenyl]-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 823.06
Molecular Formula: C60H46N4
InChi Key: XOYZGLGJSAZOAG-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results