accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 2100-31-4

IUPAC Name: 2-propoxybenzoate
Synonyms: 2-propoxybenzoate
Molecular Weight (g/mol): 179.2
Molecular Formula: C10H11O3
InChi Key: OXOWWPXTTOCKKU-UHFFFAOYSA-M
SMILES: CCCOC1=CC=CC=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results