accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2100-42-7

IUPAC Name: 2-chloro-1,4-dimethoxybenzene
Synonyms: 1,4-dimethoxy-2-chlorobenzene
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: QMXZSRVFIWACJH-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)