accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOS

CAS RN 21010-06-0

IUPAC Name: 5-(thiophen-2-yl)pentanoic acid
Synonyms: 5-(thiophen-2-yl)pentanoic acid
Molecular Weight (g/mol): 184.25
Molecular Formula: C9H12O2S
InChi Key: OQFTZRHAQGXEQU-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results