accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 210169-57-6

CH3POOOOPCH3CH3CH3H3CH3CCH3CH3(?)

CAS RN 210169-57-6

IUPAC Name: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane
Synonyms: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 722.8
Molecular Formula: C46H44O4P2
InChi Key: XJJVPYMFHXMROQ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)