accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH3COCH3

CAS RN 2103-57-3

IUPAC Name: 2,3,4-trimethoxybenzaldehyde
Synonyms: 2,3,4-trimethoxybenzaldehyde
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: UCTUXUGXIFRVGX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(OC)=C1OC
Boiling Point 
  • (4)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)