CAS RN 210344-95-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Caspase-3 Inhibitor II
The Caspase-3 Inhibitor II, also referenced under CAS 210344-95-9, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.