accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSNNHH2N

CAS RN 21036-64-6

IUPAC Name: 4-(furan-2-yl)-2-hydrazinyl-1,3-thiazole
Synonyms: 4-(furan-2-yl)-2-hydrazinyl-1,3-thiazole
Molecular Weight (g/mol): 181.21
Molecular Formula: C7H7N3OS
InChi Key: CHVBUBDKTCFMMR-UHFFFAOYSA-N
SMILES: NNC1=NC(=CS1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results