accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNS

CAS RN 21036-67-9

IUPAC Name: 2-methyl-4-(thiophen-2-yl)-1,3-thiazole
Synonyms: 2-methyl-4-(thiophen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 181.27
Molecular Formula: C8H7NS2
InChi Key: HILMKDNBZMYHDJ-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results