accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21036-68-0

IUPAC Name: 2-hydrazinyl-4-(thiophen-2-yl)-1,3-thiazole
Synonyms: 2-hydrazinyl-4-(thiophen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 197.27
Molecular Formula: C7H7N3S2
InChi Key: QYFFQQHJTLJVGU-UHFFFAOYSA-N
SMILES: NNC1=NC(=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results