accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 2104-19-0

IUPAC Name: 9-methoxy-9-oxononanoic acid
Synonyms: 9-methoxy-9-oxononanoic acid
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: VVWPSAPZUZXYCM-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCC(O)=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results