accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOFFF

CAS RN 2105-61-5

IUPAC Name: 1,2,4-trifluoro-5-nitrobenzene
Synonyms: 1,2,4-trifluoro-5-nitrobenzene
Molecular Weight (g/mol): 177.08
Molecular Formula: C6H2F3NO2
InChi Key: ROJNMGYMBLNTPK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(F)C=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results