accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClPNP

CAS RN 21050-13-5

IUPAC Name: bis(triphenyl-λ⁵-phosphanylidene)azanium chloride
Synonyms: bis(triphenylphosphine)iminium chloride
Molecular Weight (g/mol): 574.04
Molecular Formula: C36H30ClNP2
InChi Key: LVRCYPYRKNAAMX-UHFFFAOYSA-M
SMILES: [Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results