accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2106-49-2

IUPAC Name: 1-chloro-2-fluoro-3-nitrobenzene
Synonyms: 1-chloro-2-fluoro-3-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: RBAHXNSORRGCQA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results