accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21062-20-4

IUPAC Name: 2-(2-chloro-2-oxoethoxy)acetyl chloride
Synonyms: 2-(2-chloro-2-oxoethoxy)acetyl chloride
Molecular Weight (g/mol): 170.97
Molecular Formula: C4H4Cl2O3
InChi Key: GTZXSBQCNBNWPK-UHFFFAOYSA-N
SMILES: ClC(=O)COCC(Cl)=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)