accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBr

CAS RN 210705-84-3

IUPAC Name: 2-bromo-4-hexylthiophene
Synonyms: 2-bromo-4-hexylthiophene
Molecular Weight (g/mol): 247.19
Molecular Formula: C10H15BrS
InChi Key: IKXKKFNSZQNRFR-UHFFFAOYSA-N
SMILES: CCCCCCC1=CSC(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results