accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOSOOOCH3SOOOCH3SCH3CH3H3CSOOOCH3CH3CH3CH3CH3CH3H3CH3CH3CH3C

CAS RN 210706-03-9

IUPAC Name: ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate
Synonyms: ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate
Molecular Weight (g/mol): 1065.42
Molecular Formula: C56H72O12S4
InChi Key: IGVIIKFHGZNQFL-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=C2SC3=CC(=CC(SC4=C(OCC(=O)OCC)C(SC5=C(OCC(=O)OCC)C(SC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results