accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 210820-99-8

H3CH3CCH3OONNOOHNHOONH2

CAS RN 210820-99-8

IUPAC Name: 3-{1-[(tert-butoxy)carbonyl]-1H-imidazol-4-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid; cyclohexanamine
Synonyms: 3-[1-(tert-butoxycarbonyl)imidazol-4-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid; cyclohexylamine
Molecular Weight (g/mol): 576.69
Molecular Formula: C32H40N4O6
InChi Key: UDEXYAHZNOKVIG-UHFFFAOYNA-N
SMILES: NC1CCCCC1.CC(C)(C)OC(=O)N1C=NC(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)