accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3CH3HO

CAS RN 2110-78-3

IUPAC Name: methyl 2-hydroxy-2-methylpropanoate
Synonyms: methyl 2-hydroxy-2-methylpropanoate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: XYVQFUJDGOBPQI-UHFFFAOYSA-N
SMILES: COC(=O)C(C)(C)O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)